Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRES17 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987189
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7189 Supplier Bioss Supplier No. BS6299R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phosphodiesterase (PDE) that has higher activity toward cAMP than cGMP, as substrate. It plays a role in cell proliferation, is able to induce cell motility and acts as a negative regulator of NME1.

Specifications

Antigen DRES17
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PRUNE1
Gene Accession No. Q86TP1
Gene Alias 9230112O05Rik; C130058A12; DRES17; DRES-17; Drosophila-related expressed sequence 17; Exopolyphosphatase PRUNE1; hPrune; H-PRUNE; HTcD37; protein prune homolog; Protein prune homolog 1; Prune; prune exopolyphosphatase; prune homolog; prune homolog (Drosophila); Prune1; PRUNEM1; Prune-M1
Gene Symbols PRUNE1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DRES17.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58497
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.