Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRG1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | DRG1 |
---|---|
Concentration | 0.05mg/mL |
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
DRG1 Polyclonal specifically detects DRG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DRG1 | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
developmentally regulated GTP binding protein 1, developmentally regulated GTP-binding protein 1, developmentally-regulated GTP-binding protein 1, DKFZp434N1827, DRG-1, NEDD3, NEDD-3, Neural precursor cell expressed developmentally down-regulated protein 3, neural precursor cell expressed, developmentally down-regulated 3 | |
DRG1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
0.05mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
4733 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ELEGFGIRLNSKPPNIGFKKKDKGGINLTATCPQSELDAETVKSILAEYKIHNADVTLRSDATADDLIDVVEGNRVYIPCIYVLNKIDQISIEELDIIYKVPHCVPISAHHRWNFDDLLEKIWDYLKLVRIYTKPKGQLPDYT | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title