Learn More
Invitrogen™ DRG1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total DRG1 protein.
Specifications
Specifications
| Antigen | DRG1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | DRG1 |
| Gene Accession No. | P32233, Q9Y295 |
| Gene Alias | AA408859; AI132520; developmentally regulated GTP binding protein 1; developmentally regulated GTP-binding protein 1; developmentally-regulated GTP-binding protein 1; Drg; Drg1; DRG-1; GTP-binding protein; NEDD3; NEDD-3; neural precursor cell expressed developmentally down-regulated protein 3; neural precursor cell expressed, developmentally down-regulated 3; TRAFAC GTPase DRG1; Translation factor GTPase DRG1 |
| Gene Symbols | DRG1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.