Learn More
DRG1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305384AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
May play a role in cell proliferation, differentiation and death.Specifications
DRG1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P32233, Q9Y295 | |
Drg1 | |
Between 200 and 250 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
DRG1 | |
Developmentally-regulated GTP-binding protein 1, DRG-1, Neural precursor cell expressed developmentally down-regulated protein 3, NEDD-3, DRG1, NEDD3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13494, 4733 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.