Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216227
Description
DRG2 Polyclonal specifically detects DRG2 in Human, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DRG2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
developmentally regulated GTP binding protein 2, developmentally regulated GTP-binding protein 2, developmentally-regulated GTP-binding protein 2, DRG-2 | |
Rabbit | |
41 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (96%), Chicken (96%), Xenopus laevis (94%), Zebrafish (92%), Bovine (98%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
P55039 | |
DRG2 | |
Recombinant protein encompassing a sequence within the center region of human DRG2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
1819 | |
Human, Xenopus species | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction