Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRG2 Polyclonal Antibody, Invitrogen™

Catalog No. PA549155
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549155 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549155 Supplier Thermo Scientific Supplier No. PA549155

Rabbit Polyclonal Antibody

Predicted to react with bovine based on sequence homology.

3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRG2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Drg2
Gene Accession No. P55039, Q9QXB9
Gene Alias AI255295; AI461605; developmentally regulated GTP binding protein 2; developmentally regulated GTP-binding protein 2; developmentally-regulated GTP-binding protein 2; DRG2; DRG-2; RGD1562380; TRAFAC GTPase DRG2; Translation factor GTPase DRG2
Gene Symbols Drg2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 154-180 amino acids from the Central region of human DRG2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13495, 1819
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.