Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRGX Antibody, mFluor Violet 450 SE, Novus Biologicals™

Chicken Polyclonal Antibody
Supplier: Novus Biologicals NBP241138MFV450
Description
DRGX Polyclonal antibody specifically detects DRGX in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| DRGX | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| dorsal root ganglia homeobox, DRG11, paired related homeobox-like 1, paired-like homeodomain trancription factor DRG11, paired-related homeobox protein-like 1, PRRXL1 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human DRGX. The immunogen is located within the last 50 amino acids of DRGX. Amino Acid Squence: VAKPAPPDGSQEKTSPTKE | |
| 0.1 mL | |
| Neuroscience | |
| 644168 | |
| Store at 4C in the dark. | |
| IgY |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction