Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DRGX Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548376

Catalog No. PA548376


Only null left
Add to Cart

Description

Description

DRGX Polyclonal Antibody for Western Blot

Transcription factor required for the formation of correct projections from nociceptive sensory neurons to the dorsal horn of the spinal cord and normal perception of pain.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DRGX
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
A6NNA5, Q8BYH0
DRGX, Prrxl1
KLH conjugated synthetic peptide between 234-263 amino acids from the C-terminal region of human DRGX.
400 μL
Primary
Human, Mouse
Antibody
IgG
Western Blot
Unconjugated
DRGX
dorsal root ganglia homeobox; dorsal root ganglia homeobox protein; Dorsal root ganglion 11; drg11; Drgx; Homeobox protein DRG11; paired homeodomain protein DRG11; paired related homeobox protein-like 1; paired related homeobox protein-like 1b; paired related homeobox-like 1; paired-like homeodomain trancription factor DRG11; paired-like homeodomain transcription factor DRG11; paired-related homeobox protein-like 1; Prrxl1
Rabbit
Antigen affinity chromatography, Protein A
RUO
107751, 644168
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.