Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DRGX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560570
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560570 Supplier Invitrogen™ Supplier No. PA560570
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PMGLSFLPTY GCQSNRTASV ATLRMKAREH SEAVLQSANL LPSTSSSPGP VAKPAPPDGS QEKTSPTKEQ SEAEKSV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 88%.

Transcription factor required for the formation of correct projections from nociceptive sensory neurons to the dorsal horn of the spinal cord and normal perception of pain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRGX
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DRGX
Gene Accession No. A6NNA5
Gene Alias dorsal root ganglia homeobox; dorsal root ganglia homeobox protein; Dorsal root ganglion 11; drg11; Drgx; Homeobox protein DRG11; paired homeodomain protein DRG11; paired related homeobox protein-like 1; paired related homeobox protein-like 1b; paired related homeobox-like 1; paired-like homeodomain trancription factor DRG11; paired-like homeodomain transcription factor DRG11; paired-related homeobox protein-like 1; Prrxl1
Gene Symbols DRGX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DRGX. Recombinant protein control fragment (Product #RP-99872).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644168
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.