Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DRGX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569486
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569486 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569486 Supplier Invitrogen™ Supplier No. PA569486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Yeast: 79%; Zebrafish: 100%.

Transcription factor required for the formation of correct projections from nociceptive sensory neurons to the dorsal horn of the spinal cord and normal perception of pain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRGX
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DRGX
Gene Accession No. A6NNA5
Gene Alias dorsal root ganglia homeobox; dorsal root ganglia homeobox protein; Dorsal root ganglion 11; drg11; Drgx; Homeobox protein DRG11; paired homeodomain protein DRG11; paired related homeobox protein-like 1; paired related homeobox protein-like 1b; paired related homeobox-like 1; paired-like homeodomain trancription factor DRG11; paired-like homeodomain transcription factor DRG11; paired-related homeobox protein-like 1; Prrxl1
Gene Symbols DRGX
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human DRGX.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644168
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.