Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Applied Biosystems™ dRhodamine Dye Terminator Ready Reaction Cycle Sequencing Kit

Click to view available options
No. of Reactions:
100 Reactions
1000 Reactions
Description
Includes
Each kit contains 800uL tube of dRhodamine terminator ready reaction mix, 1 tube M13 (-21) primer, 1 tube pGEM control DNA, protocol manual.
- Increase your signal-to-noise ratio
- Simplify workflows with easy-to-clean-up chemistries
- Get optimized chemistry for sequencing templates with homo-polymers (AA's, TT's, etc.)
- Obtain clean signal, even peak patterns, and minimal mobility shifts with improved dye terminators
- Reduce spectrum overlap with narrow dye emission spectra

Specifications
Specifications
Content And Storage | Each kit contains 800uL Tube of dRhodamine Terminator Ready Reaction Mix, 1 Tube M13 (-21) Primer, 1 Tube pGEM Control DNA, Protocol Manual. Store kit at -15 to -25°C and avoid repeated freeze-thaw cycles. |
Format | 8-well Strip, 96-well Plate |
For Use With (Equipment) | 310 Genetic Analyzer, 3130xl Genetic Analyzer, ABI PRISM™ 377, GeneAmp 2700, GeneAmp 9600, GeneAmp 9700, GeneAmp™ 2400, 3130 Genetic Analyzer |
For Use With (Application) | Standard Sequencing |
Includes | dRhodamine Dye Terminator Ready Reaction Cycle Sequencing Kit, 100 rxns, 8-well Strips, 96-well Plate, For 310 Genetic Analyzer, 3130 Genetic Analyzer |
No. of Reactions | 100 Reactions |
Product Type | Sequencing Kit |
Quantity | 100 reactions |
Technique | Fluorescent Dye Terminator Sequencing |
Template Compatibility | Single Stranded DNA, Plasmid DNA (≤15Kb), PCR Amplicons |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction