Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Amphetamine Drugs of Abuse Assays

Catalog No. p-4528173
Click to view available options
Quantity:
3 x 18 mL
1 x 100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Eliminate time-consuming steps while achieving high analytical accuracy when you use Thermo Scientific™ DRI™ Amphetamine Drugs of Abuse Assays.

Eliminate time-consuming steps while achieving high analytical accuracy when you use Thermo Scientific™ DRI™ Amphetamine Drugs of Abuse Assays—ready to use in liquid form. These assays have been field-tested on a wide range of analyzers, and they are enthusiastically accepted in laboratories where cost-control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets MGC Multi-Drug Controls
Description DRI Amphetamine Drugs of Abuse Assays
Detectable Analytes Amphetamine/Methamphetamine
DoA Calibrators DRI Multi-drug Calibrators
Sample Type Urine
Type Reagent
Quantity 1 x 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.