Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Controls
GSA_VA

Catalog No. 23278323
Click to view available options
Concentration:
300 ng/mL
700 ng/mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Reduce time-consuming reagent preparation with Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Controls—ready-to-use controls in lilquid form.

Reduce time-consuming reagent preparation with Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Controls—ready-to-use controls in lilquid form. Delivering high analytical accuracy, these assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description DRI Cotinine Drugs of Abuse Controls
Concentration 700 ng/mL
Detectable Analytes Creatinine
Sample Type Urine
Type Control
Level High (700ng/mL) and Low (300ng/mL)
Quantity 1 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.