Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Controls

Catalog No. p-4527688
Click to view available options
Concentration:
300 ng/mL
700 ng/mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Reduce time-consuming reagent preparation with Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Controls—ready-to-use controls in lilquid form.

Reduce time-consuming reagent preparation with Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Controls—ready-to-use controls in lilquid form. Delivering high analytical accuracy, these assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Description DRI Cotinine Drugs of Abuse Controls
Concentration 300 ng/mL
Detectable Analytes Creatinine
Sample Type Urine
Type Control
Quantity 1 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.