Learn More
Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Calibrators

Description
Ensure accuracy and precision of your drug testing results by partnering these liquid, ready-to-use Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Calibrators with our Thermo Scientific DRI Ethyl Glucuronide (EtG) Drugs of Abuse Assay and Control Sets. These calibrators are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.
- Specific detection of Ethyl Glucuronide (EtG)
- Qualitative or semi-quantitative modes
- Excellent correlation to LC-MS/MS
- Applications for an array of clinical chemistry analyzers
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | DRI Ethyl Glucuronide (EtG) 500 Calibrator. For Criminal Justice and Forensic Use Only. For sale in the U.S.A. only. |
| Concentration | 500 ng/mL |
| Detectable Analytes | Ethyl Glucuronide (EtG) |
| Sample Type | Urine |
| Type | Calibrator |
| Packaging | 1 Bottle |
| Quantity | 1 x 10 mL |
For sale in the U.S.A. only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.