Learn More
Thermo Scientific™ DRI™ Fentanyl II Drugs of Abuse Assay
Description
Developed for hospital laboratories, addiction treatment centers, and toxicology laboratories, the Thermo Scientific™ DRI™ Fentanyl II Drugs of Abuse Assay is an effective tool for the screening fentanyl abuse and misuse. Its ability to detect both fentanyl and its primary metabolite, norfentanyl gives this assay a broader window of detection compared to competing products. This automated urine drug screening assay exhibits negligible cross-reactivity to other opioids. It is the only 510(k) cleared assay that combines norfentanyl cross-reactivity with a low 1 ng/mL cutoff concentration.
- Specifically detect fentanyl and its major metabolite, norfentanyl, at physiologically relevant concentrations in human urine
- Low cross-reactivity to other non-fentanyl synthetic opioids and over-the-counter medication
- High sensitivity with a cutoff concentration of 1 ng/mL
- Superior assay precision
- Excellent correlation to LC-MS/MS
- Liquid, Ready-to-Use

Specifications
Specifications
Content And Storage | 2°C to 8°C |
Control Sets | DRI Fentanyl II Control Kit |
Description | DRI Fentanyl II Drugs of Abuse Assay |
Detectable Analytes | Fentanyl (FEN) |
DoA Calibrators | DRI Fentanyl II DoA Assay |
Type | Reagent |
Quantity | 1 x 500 mL |
For sale in the U.S.A. and Canada only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.