Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Assay

Catalog No. 23278376 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3 x 18 mL
1 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-376 3 x 18 mL
21-200-422 1 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-278-376 Supplier Thermo Scientific™ Supplier No. 10018054
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Save time while maintaining accuracy with the liquid, ready-to-use Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Assay.

Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of the Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Assay, which offers applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs. This assay is for sale in the U.S.A. and Canada only. Supports new SAMHSA guidelines (effective October 1, 2017)

  • Specific detection of hydrocodone
  • Qualitative or semi-quantitative modes
  • Excellent correlation to LC-MS/MS
  • Applications for an array of clinical chemistry analyzers
  • For sale in the U.S.A. and Canada only


TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets DRI Hydrocodone Control Set
Description DRI Hydrocodone Drugs of Abuse Assay for Indiko
Detectable Analytes Hydrocodone
DoA Calibrators Hydrocodone Calibrator, Negative Urine Calibrator
Sample Type Urine
Type Reagent
Quantity 3 x 18 mL

Disclaimers: Some of these products are not cleared in every country. Please contact your sales representative for more information. Product claims may differ from country to country based on regulations and product approvals. Please contact your country representative for further details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.