Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ DRI™ Propoxyphene Drugs of Abuse Assay
Click to view available options
Quantity:
3 x 18 mL
1 x 100 mL
Description
Eliminate time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Propoxyphene Drugs of Abuse Assay. This liquid, ready-to-use assay delivers high analytical accuracy for an array of clinical chemistry analyzers. This liquid, ready-to-use assay has been field-tested on a wide range of analyzers. It is enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Control Sets | MGC Primary DAU Control Set |
| Description | DRI Propoxyphene Drugs of Abuse Assay |
| Detectable Analytes | Propoxyphene |
| DoA Calibrators | DRI Multi-drug Calibrators |
| Sample Type | Urine |
| Type | Reagent |
| Quantity | 1 x 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction