Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Calibrators

Catalog No. p-4527616
Click to view available options
Quantity:
1 x 5 mL
1 x 25 mL
Concentration:
100 ng/mL
20 ng/mL
200 ng/mL
50 ng/mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Utilize the globally valued accuracy, precision and lot-to-lot dependability of DRI THC(Cannabinoid) Drugs of Abuse Calibrators.

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ THC(Cannabinoid) Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI; 50ng/mL; 5mL1
Concentration 50 ng/mL
Detectable Analytes 11-nor-9-COOH-D9-THC
Sample Type Urine
Type Calibrator
Packaging 1 Bottle
Quantity 1 x 5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.