Learn More
Thermo Scientific™ DRI Tricyclics Serum Toxicology Assays
Description
Quickly and accurately identify tricyclics in serum samples with Thermo Scientific™ DRI™ Tricyclics Serum Toxicology Assays, excellent tools for emergency rooms when urine specimens are not conveniently available.
These Serum Toxicology Tricyclics Assays have the flexibility of analyzing serum or urine samples. These assays are provided in convenient packaging with separate calibrators for ease of use. A multi-analyte Serum Tox Control is also available. The assays are liquid, ready-to-use and have the versatility to be run on a variety of clinical chemistry analyzers.
- Serum, plasma-and urine-based assay
- Used to identify class of drug overdose
Specifications
Specifications
| Description | DRI Tricyclics Serum Toxicology Assays |
| Detectable Analytes | Tricyclic Antidepressants |
| Sample Type | Serum, Plasma, Urine |
| Type | Reagent |
| Quantity | 1 X 500 mL |
For sale in the U.S.A. and Canada only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.