Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Drierite™, with indicator, 10-20 mesh
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging:
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
Specifications
Specifications
| Color | Blue |
| Melting Point | 1450.0°C |
| Physical Form | Irregular Granules |
| Quantity | 10 kg |
| UN Number | 3077 |
| Packaging | Plastic Drum |
| Solubility Information | Solubility in water: slightly soluble. Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents. SiO2 and insolubles: 0.58% |
| R2O | 0.29% |
| IUPAC Name | calcium sulfate |
| Formula Weight | 136.13 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing genetic defects. May cause cancer by inhalation. May damage fertility. Causes serious eye irritation. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF exposed or concerned: Get medical advice/attention. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction