Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRIFIRE™ Flight Deck Jerseys
SDP

Catalog No. 17001105 Shop All National Public Seating Corporation Products
Click to view available options
Color:
Blue
Brown
Green
Off White
Purple
Red
Yellow
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Provide easy visual identification on shipboard flight decks

  • Lightweight DRIFIRE fabric dries four times faster that traditional cotton
  • Inherently flame-resistant — will not melt or fuse to skin when exposed to open flame
  • Mock turtleneck design increases coverage and protection
  • Thumbholes ensure sleeves stay in place
  • Excellent colorfastness — withstands standard shipboard laundry
  • Resists oil stains
  • Controls odor
  • Superior moisture wicking helps regulate skin temperature

Applications:

Industry: Navy.

Specifications

Size 3X-Large
Material Lightweight DRIFIRE fabric
Color Off White
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.