Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ DH Air Vented Work Shirt
SDP

Catalog No. 17010909 Shop All National Public Seating Corporation Products
Click to view available options
Size:
Medium
Large
X-Large
3X-Large
5X-Large

Catalog No. 17-010-909

Supplier: National Safety Apparel™ SHRA2TGV3XRG

Only null left
Add to Cart
Add to Cart

Work shirt features a patented vent design that actively allows hot air to exit the garment creating breathability. Covered back mesh panel wicks away sweat increasing mobility and performance.

  • Patented vent design with industry leading comfort
  • Lightweight and breathable fabric
  • Button down collar
  • Two front chest pockets with flaps and button closure
  • Quickly identify compliance with exterior labeling
  • Dual hazard protection against arc flash and flash fire

Specifications

Size 3X-Large
Gender Unisex
Material Aramid, Lyocell, Modacrylic
Color Medium Blue
Neck Style Collar
Sleeve Style Long Sleeve
Closure Type Button
Disposable Reusable
Product Type Work Shirt
Hazard Risk Category CAT 2
Arc Flash Rating 9.1 cal/cm²
Certifications/Compliance NFPA 70E, NFPA 2112, ASTM F1506, CSA Z462, OSHA 1910.269
For Use With (Application) Utilities, Oil, Gas
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.