Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRIFIRE™ Two-Piece Navy Flight Suits
SDP

Catalog No. 17000343 Shop All National Public Seating Corporation Products
Click to view available options
Closure Type:
Zipper
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed for Naval and Marine Aviators with end-user input

  • Permanent, inherent flame resistance that won't wash or wear out
  • Controls odor
  • Increased durability and FR performance
  • Unique design features to improve fit, increase range of motion and reduce snag hazards
  • Navy and USMC “Safe to Fly”
  • Meets NAVAIR specifications
  • Available in khaki color in different sizes ranging from X-Small to XXX-Large

Two-Piece Navy Flight Suit Jackets:

  • Angled shoulder pockets for easy access
  • Adjustable waist tabs for customized torso fit

US Navy, US Coast Guard, US Marines and MRO (Maintence Repair Operations)

Specifications

Size Large
Material Modacrylic, Cotton
Color Khaki
Closure Type Zipper
Resistance Flame
Certifications/Compliance NAVAIR
Description Modacrylic, Cotton, Para Aramid™, Anti-stat
No. of Pockets 4
Size (English) Chest 41 to 45 in.
For Use With (Application) Naval and marine aviators with end-user input
Length (English) 30.25 to 33.12 in.
Length (English) Sleeve 26 to 28 in.
Length (Metric) 76.8 to 84.1 cm
Length (Metric) Sleeve 66 to 71.1 cm
Height (English) 5.25 to 6.25 ft.
Height (Metric) 160 to 190.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.