Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRIFIRE™ Two-Piece Navy Flight Suits
Shop All National Public Seating Corporation Products

Click to view available options
Closure Type:
Zipper
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Permanent, inherent flame resistance that won't wash or wear out
- Controls odor
- Increased durability and FR performance
- Unique design features to improve fit, increase range of motion and reduce snag hazards
- Navy and USMC “Safe to Fly”
- Meets NAVAIR specifications
- Available in khaki color in different sizes ranging from X-Small to XXX-Large
Two-Piece Navy Flight Suit Jackets:
- Angled shoulder pockets for easy access
- Adjustable waist tabs for customized torso fit
US Navy, US Coast Guard, US Marines and MRO (Maintence Repair Operations)
Specifications
Specifications
| Size | Medium |
| Material | Modacrylic, Cotton |
| Color | Khaki |
| Closure Type | Zipper |
| Resistance | Flame |
| Certifications/Compliance | NAVAIR |
| Description | Modacrylic, Cotton, Para Aramid™, Anti-stat |
| No. of Pockets | 4 |
| Size (English) Chest | 37 to 41 in. |
| For Use With (Application) | Naval and marine aviators with end-user input |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction