Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRIL1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298579AF700
Description
DRIL1 Polyclonal specifically detects DRIL1 in Human samples. It is validated for Western Blot.Specifications
DRIL1 | |
Polyclonal | |
Western Blot | |
ARID domain-containing 3A, ARID domain-containing protein 3A, AT rich interactive domain 3A (BRIGHT- like), AT rich interactive domain 3A (BRIGHT- like) protein, AT rich interactive domain 3A (BRIGHT-like), AT-rich interactive domain-containing protein 3A, B-cell regulator of IgH transcription, Bright, dead ringer-like 1, Dead ringer-like protein 1, DRIL1dead ringer-like 1 (Drosophila), DRIL3, DRX, E2F-binding protein 1, E2FBP1 | |
Produced in rabbits immunized with E. coli-derived Human DRIL1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
1820 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction