Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRIL1/ARID3A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982555
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2555 Supplier Bioss Supplier No. BS13031R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The mouse Bright (B-cell regulator of IgH transcription) protein is expressed specifically in B cells and transactivates an IgH enhancer in transient transfection assays. Bright and the Drosophila ′dead ringer′ (dri) gene product belong to a protein family whose members share a conserved DNA-binding domain termed the ′A/T-rich interaction domain′ (ARID).

Specifications

Antigen DRIL1/ARID3A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ARID3A
Gene Accession No. Q99856
Gene Alias ARID domain-containing 3A; ARID domain-containing protein 3A; ARID3A; AT rich interactive domain 3A (Bright like); AT rich interactive domain 3A (BRIGHT- like) protein; AT rich interactive domain 3A (BRIGHT-like); AT-rich interaction domain 3A; AT-rich interactive domain-containing protein 3A; B-cell regulator of IgH transcription; Bright; dead ringer homolog 1; dead ringer-like 1; Dead ringer-like protein 1; Dri1; Dril1; DRIL3; DRX; E2F-binding protein 1; E2FBP1
Gene Symbols ARID3A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DRIL1/ARID3A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1820
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.