Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DRIL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530009
Description
Recommended positive controls: HCT116. Predicted reactivity: Mouse (92%), Rat (92%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The mouse Bright (B-cell regulator of IgH transcription) protein is expressed specifically in B cells and transactivates an IgH enhancer in transient transfection assays. Bright and the Drosophila 'dead ringer' (dri) gene product belong to a protein family whose members share a conserved DNA-binding domain termed the 'A/T-rich interaction domain' (ARID).
Specifications
| DRIL1 | |
| Polyclonal | |
| Unconjugated | |
| ARID3A | |
| ARID domain-containing 3A; ARID domain-containing protein 3A; ARID3A; AT rich interactive domain 3A (Bright like); AT rich interactive domain 3A (BRIGHT- like) protein; AT rich interactive domain 3A (BRIGHT-like); AT-rich interaction domain 3A; AT-rich interactive domain-containing protein 3A; B-cell regulator of IgH transcription; BRIGHT; dead ringer homolog 1; dead ringer-like 1; dead ringer-like protein 1; Dri1; DRIL1; DRIL3; DRX; E2F-binding protein 1; E2FBP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13496, 1820 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.69 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q62431, Q99856 | |
| ARID3A | |
| Recombinant protein encompassing a sequence within the C-terminus region of human DRIL1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction