Learn More
Stoelting™ Drill Bit
SDP
Stoelting™ Carbide Drill Bits cut cleaner and retain sharp edges longer.
$65.00
Specifications
For Use With (Application) Neuroscience
Material Carbide
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
10-001-086
SDP
View Documents
Stoelting™
514552
0.60 mm
Pack of 6 for $65.00
Only null left
Add to Cart
10-001-087
SDP
View Documents
Stoelting™
514553
0.75 mm
Pack of 6 for $65.00
Only null left
Add to Cart
10-001-088
SDP
View Documents
Stoelting™
514554
1 mm
Pack of 6 for $65.00
Only null left
Add to Cart
10-001-089
SDP
View Documents
Stoelting™
514555
1.35 mm
Pack of 6 for $65.00
Only null left
Add to Cart
10-001-090
SDP
View Documents
Stoelting™
514556
1.75 mm
Pack of 6 for $65.00
Only null left
Add to Cart
Description
Description
  • Generally used in Stereotaxic Surgery and procedures
  • Suitable for use with the 58650 Flexible Shaft Drill, and the 51449 Micromotor Drill
  • Spherical heads, variety of sizes
  • Packages of 6 bits (all the same size)
Specifications
Specifications
Neuroscience
Carbide
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.