Learn More
Stoelting™ Drill and Injection Robot for Motorized Stereotaxic Instruments
SDP
Provides for a fully software driven solution to stereotaxic surgery
Supplier:  Stoelting™ 58643
 View more versions of this product
Catalog No. 10-000-275
Only null left
Add to Cart
Request a Quote
Description
Description
  • Allows for high precision injection
  • After loading the syringe, the injection robot allows for multiple injections in different animals without tool exchange or recalibration
  • Integrated into the StereoDrive software (computer and motorized stereotaxic not included but required)
  • Available as Glass-Capillary Nanoinjector
  • Atlas Integration
  • High Throughput Drilling and Injection
  • No Tool Exchange
  • Defined/Stored Protocols
  • Multisite Microinjections
  • Ideal for Optogenetics applications
  • Precise Depth Control
  • Brain Windowing
  • Contour Drilling
  • Smooth Edges
  • Skull Thinning
Specifications
Specifications
Neuroscience
Robot Drill and Injection Bundle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.