Learn More
Stoelting™ Drill and Injection Robot for Motorized Stereotaxic Instruments
SDP
Provides for a fully software driven solution to stereotaxic surgery
$4560.00 - $27500.00
Specifications
For Use With (Application) Neuroscience
View More Specs
Products 4
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-000-201
SDP
View Documents
Stoelting™
51722
Laptop Computer, Stereotaxic System
Each for $4,560.00
Only null left
Add to Cart
10-000-277
SDP
View Documents
Stoelting™
58645
Injection Robot
Each for $14,300.00
Only null left
Add to Cart
10-000-275
SDP
View Documents
Stoelting™
58643
Robot Drill and Injection Bundle
Each for $27,500.00
Only null left
Add to Cart
10-000-274
SDP
View Documents
Stoelting™
58640
Robot Drill
Each for $14,300.00
Only null left
Add to Cart
Description
Description
  • Allows for high precision injection
  • After loading the syringe, the injection robot allows for multiple injections in different animals without tool exchange or recalibration
  • Integrated into the StereoDrive software (computer and motorized stereotaxic not included but required)
  • Available as Glass-Capillary Nanoinjector
  • Atlas Integration
  • High Throughput Drilling and Injection
  • No Tool Exchange
  • Defined/Stored Protocols
  • Multisite Microinjections
  • Ideal for Optogenetics applications
  • Precise Depth Control
  • Brain Windowing
  • Contour Drilling
  • Smooth Edges
  • Skull Thinning
Specifications
Specifications
Neuroscience
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.