Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRIP130 Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502207759
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7759 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7759 Supplier Fabgennix Inc Supplier No. MED23101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DRIP130 Polyclonal antibody specifically detects DRIP130 in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen DRIP130
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene MED23
Gene Accession No. Q5EB59, Q80YQ2, Q9ULK4
Gene Alias 130 kDa transcriptional co-activator; 130kDa; 133 kDa transcriptional co-activator; 3000002A17Rik; activator-recruited cofactor 130 kDa component; ARC130; Cofactor required for Sp1 transcriptional activation subunit 3; cofactor required for Sp1 transcriptional activation, subunit 3 (130kD); CRSP complex subunit 3; CRSP130; CRSP133; Crsp3; DRIP130; ESTM7; hSur-2; KIAA1216; Med23; Mediator complex subunit 23; mediator complex subunit MED23 variant MED23_i6; mediator complex subunit MED23 variant MED23_i7; mediator of RNA polymerase II transcription subunit 23; mKIAA1216; MRT18; mSur-2; Protein sur-2 homolog; SUR2; SUR-2; Transcriptional coactivator CRSP130; vitamin D3 receptor interacting protein; Vitamin D3 receptor-interacting protein complex 130 kDa component; X83317
Host Species Rabbit
Immunogen Synthetic pepetide taken within amino acid region 650-700 on MED23 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309565, 472124, 70208, 9439
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.