Learn More
Cytiva™ Drive Sleeve for Chromatography System
Catalog No. 01257019 Shop All Cytiva Products
Change view
Click to view available options
Packaging Quantity:
5 Sleeves
Catalog No. Packaging Quantity
01-257-019 5 Sleeves
1 options
This item is not returnable. View return policy
Catalog No. 01-257-019 Supplier Cytiva™ Supplier No. 19606702
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Drive sleeves for use with chromatography systems.
  • Place these sleeves on the drive bushing to prevent bowl slippage
TRUSTED_SUSTAINABILITY
Specifications
Type Sleeve
Color Black
Packaging Quantity 5 Sleeves