Learn More
Sigma Aldrich Droplet Generator Chip - Multi Channel Design Fluidic 440 Pc
Catalog No. 9215051EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. 92-150-51EA
Supplier: Sigma Aldrich 9215051EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Microfluidic generation of droplets can produce highly monodispersed droplets with high frequency (up to hundreds of kHz). Interest in droplet-based microfluidic systems has grown substantially, because microfluidics offers the ability to handle very small volumes (mul to fl) of fluids, provides better mixing, encapsulation, sorting, and sensing. Microfluidics can be used for high throughput experimentation. Microfluidic-based droplets have many diverse and varied applications such as particle synthesis and chemical analysis. Highly controlled droplet production also makes single cell analysis, or drug testing possible.Droplet generator chip - Multi channel design, Fluidic 440, PC is made of PC (polycarbonate) is suited to evaluate droplet generation with a single cross, flow focusing geometry. The nozzle sizes are 50, 60, 70 and 80 mum on this chip. With its two Mini Luer inlet and one Mini Luer outlet ports per droplet generation unit, the chip requires a two-channel microfluidic pump. There are 8 droplet generator units on this chip.
Specifications
Quantity 1 Ea.
Synonym Microfluidic; Microparticle; Nanoparticle
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.