Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drosha Antibody, Novus Biologicals™
SDP

Catalog No. NBP103349 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP103349 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP103349 Supplier Novus Biologicals Supplier No. NBP103349
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

Drosha Polyclonal specifically detects Drosha in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen Drosha
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2-5 mcg/mg lysate, Immunohistochemistry-Frozen
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q9NRR4
Gene Alias drosha, double-stranded RNA-specific endoribonuclease, drosha, ribonuclease type III, EC 3.1.26.3, Etohi2, nuclear RNase III Drosha, p241, Protein Drosha, putative protein p241 which interacts with transcription factor Sp1, putative ribonuclease III, RANSE3L, ribonuclease 3, Ribonuclease III, ribonuclease III, nuclear, RN3nuclear, RNase III
Gene Symbols DROSHA
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1324 and 1374 of human double-stranded RNA-specific endoribonuclease using the numbering given in entry NP_037367.3 (GeneID 29102).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics, microRNA
Primary or Secondary Primary
Gene ID (Entrez) 29102
Test Specificity Mouse reactivity reported in scientific literature (PMID: 25351346) Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee, African elephant and Thirteen-lined ground squirrel.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.