Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drosha Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572012
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2012 10 μL
50-157-2011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2012 Supplier Bethyl Laboratories, Inc Supplier No. IHC00389T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Members of the ribonuclease III superfamily of double-stranded RNA-specific endoribonucleases participate in diverse RNA maturation and decay pathways in eukaryotic and prokaryotic cells processing in the nucleus .

Specifications

Antigen Drosha
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene DROSHA
Gene Accession No. Q5HZJ0, Q9NRR4
Gene Alias OTTHUMP00000220910; OTTHUMP00000220911; OTTHUMP00000220912; OTTHUMP00000220913; RNase III; drosha, double-stranded RNA-specific endoribonuclease; drosha, ribonuclease type III; nuclear RNase III Drosha; p241; putative protein p241 which interacts with transcription factor Sp1; putative ribonuclease III; ribonuclease 3; ribonuclease III, nuclear
Gene Symbols DROSHA
Host Species Rabbit
Immunogen Between 1050 and 1100
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14000, 29102
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.