Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drosha Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501735922 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5922 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5922 Supplier Proteintech Group Inc Supplier No. 550011AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody is specific to DROSHA. DROSHA/RNASEN has some transcription forms with MW 160 kDa, 151 kDa, 138 kDa, and 90 kDa.

Polyglutamylase which preferentially modifies beta-tubulin and nucleosome assembly proteins NAP1 and NAP2. Involved in the side-chain initiation step of the polyglutamylation reaction rather than in the elongation step.

Specifications

Antigen Drosha
Applications Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DROSHA
Gene Accession No. Q5HZJ0, Q9NRR4
Gene Alias DROSHA, ETOHI2, HSA242976, p241, RANSE3L, Ribonuclease 3, Ribonuclease III, ribonuclease type III, nuclear, RN3, RNase III, RNASE3L, RNASEN
Gene Symbols Drosha
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14000, 29102, 310159
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.