Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Drosophila Smad2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7948
This item is not returnable.
View return policy
Description
Smad2 Polyclonal specifically detects Smad2 in Drosophila samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Smad2 | |
Polyclonal | |
Unconjugated | |
O96660 | |
SMAD2 | |
E. coli-derived recombinant Drosophila Smad2 Gly262-Ser486 Accession # O96660 | |
100 μg | |
Primary | |
Detects Drosophila Smad2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) Smad1 is observed, and less than 1% cross-reactivity with rhSmad2, rhSmad3, and rhSmad5 is observed. | |
Drosophila | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1-0.7 ug/mL, Immunohistochemistry 0.1-0.7 ug/mL | |
hMAD-2, hSMAD2, JV18, JV18-1, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2, MADH2MGC34440, MADR2, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2, Smox | |
Sheep | |
Affinity Purified | |
RUO | |
4087 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction