Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRP1 Antibody (4E11B11), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280702H
Description
DRP1 Monoclonal antibody specifically detects DRP1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
DRP1 | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
4E11B11 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
DLP1, DRP1Dnm1p/Vps1p-like protein, DVLPEC 3.6.5.5, Dymple, DYMPLEDYNIV-11, dynamin 1-like, Dynamin family member proline-rich carboxyl-terminal domain less, dynamin-1-like protein, Dynamin-like protein, Dynamin-like protein 4, Dynamin-like protein IV, Dynamin-related protein 1, FLJ41912, HDYNIV, VPS1 | |
Partial recombinant human DRP1 (between residues 100-250) expressed in E. coli. [Uniprot: O00429] | |
0.1 mL | |
Apoptosis, Cell Biology, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Signal Transduction | |
10059 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction