Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRP1 Antibody (4E11B11), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$206.50 - $499.50

Specifications

Antigen DRP1
Clone 4E11B11
Concentration 1.0 mg/mL
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP223489SS
SDP
View Documents
Novus Biologicals
NBP223489SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP223489
SDP
View Documents
Novus Biologicals
NBP223489
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

DRP1 Monoclonal specifically detects DRP1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

DRP1
1.0 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Apoptosis, Cell Biology, Mitochondrial Fusion Proteins, Neurodegeneration, Neuroscience, Signal Transduction
O00429
10059
Partial recombinant human DRP1 (between residues 100-250) expressed in E. coli. [Uniprot: O00429]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
4E11B11
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Monoclonal
Precipitation
RUO
Human, Rat
DLP1, DRP1Dnm1p/Vps1p-like protein, DVLPEC 3.6.5.5, Dymple, DYMPLEDYNIV-11, dynamin 1-like, Dynamin family member proline-rich carboxyl-terminal domain less, dynamin-1-like protein, Dynamin-like protein, Dynamin-like protein 4, Dynamin-like protein IV, Dynamin-related protein 1, FLJ41912, HDYNIV, VPS1
DNM1L
IgG1
Ammonium sulfate precipitation
Human. Expected to react with Rat, Mouse, Bovine and Chinese hamster based on sequence homology.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.