Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577924 Supplier Invitrogen™ Supplier No. PA577924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: MCF-7, mouse brain Predicted Reactivity: Rat (91%), Xenopus laevis (83%), Chicken (90%), Rhesus Monkey (98%), Bovine (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Collapsin-response mediator proteins (CRMPs) are highly expressed in the developing brain where they play major roles in axonal outgrowth, neurite differentiation, and apoptosis. Their continued expression in areas of high synaptic remodeling such as the cerebellum, hippocampus, and the olfactory system suggests that these proteins may also be involved in adult brain plasticity. CRMP-1 was initially identified as a dihydro-pyrimidinase expressed exclusively in brain; later studies have shown that it is involved with neurotrophin (NT) 3-induced neurite formation and outgrowth. CRMP-1 localization switches from axonal to somatodendritic when neurons reach functional maturity, suggesting that it is involved in early neuronal differentiation as well as in later processes related to the survival or death of the newly generated neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRMP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CRMP1
Gene Accession No. P97427, Q14194
Gene Alias Collapsin response mediator protein 1; Crmp1; CRMP-1; dihydropyrimidinase-like 1; dihydropyrimidinase-related protein 1; Dpysl1; DRP1; DRP-1; Ulip3; ULIP-3; Unc-33-like phosphoprotein 3
Gene Symbols CRMP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human CRMP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12933, 1400
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.