Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRP1 Rabbit anti-Drosophila, Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206790
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6790 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6790 Supplier Fabgennix Inc Supplier No. DRP1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DRP1 Polyclonal antibody specifically detects DRP1 in Drosophila, Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen DRP1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene DNM1L
Gene Accession No. O00429, O35303, Q8K1M6
Gene Alias 6330417M19Rik; AI450666; C-terminal region; Dlp1; dnm1l; Dnm1p/Vps1p-like protein; Dnml1; Dnmlp1; Drp1; DVLP; DYMPLE; dynamin 1 like; dynamin 1-like; dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-1-like protein-like protein; Dynamin-like protein; dynamin-like protein 1; dynamin-like protein 4; Dynamin-like protein IV; dynamin-related protein 1; DYNIV 11; DYNIV-11; EC 3.6.5.5; EGK_03514; EMPF; FLJ41912; HDYNIV; N-terminal region; python; similar to dynamin 1-like; VPS1; zgc:66163
Host Species Rabbit
Immunogen Synthetic peptide within 450-490 on human DRP1 protein
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10059, 114114, 74006
Target Species Drosophila, Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.