Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRP2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305842AF350
Description
DRP2 Polyclonal specifically detects DRP2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
DRP2 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
dystrophin related protein 2, dystrophin-related protein 2, MGC133255 | |
Produced in rabbits immunized with E. coli-derived Human DRP2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
1821 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction