Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216229
Description
DRP2 Polyclonal specifically detects DRP2 in Mouse samples. It is validated for Western Blot.Specifications
DRP2 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
dystrophin related protein 2, dystrophin-related protein 2, MGC133255 | |
Rabbit | |
108 kDa | |
0.1 mL | |
Primary | |
Mouse. Expected to cross react based on sequence identity: Human (100%), Rat (98%), Bovine (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.43mg/mL | |
Western Blot 1:500-1:3000 | |
Q13474 | |
DRP2 | |
Recombinant protein encompassing a sequence within the center region of human DRP2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
1821 | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction