Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529520
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529520 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529520 Supplier Invitrogen™ Supplier No. PA529520
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Mouse brain. Predicted reactivity: Mouse (99%), Rat (98%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Members of the dystrophin family of proteins perform a critical role in the maintenance of membrane-associated complexes at points of intercellular contact in vertebrate cells. Dystrophin related protein 2 is predicted to resemble certain short C-terminal isoforms of dystrophin and dystrophin-related protein 1 (DRP1 or utrophin). DRP2 is expressed principally in the brain and spinal cord.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DRP2
Applications Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Drp2
Gene Accession No. Q05AA6, Q13474
Gene Alias AW495265; DRP2; DRP-2; dystrophin related protein 2; dystrophin-related protein 2; dystrophin-related protein 2 A-form
Gene Symbols Drp2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DRP2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13497, 1821
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.