Learn More
Invitrogen™ DRP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551785
Description
Immunogen sequence: SRIEHFASRL AEMESQNCSF FNDSLSPDDS IDEDQYLLRH SSPITDREPA FGQQAPCSVA TESKGELQKI LAHLEDENRI LQGELRRLKW QHEEAAEAPS LADGSTEAAT DHRNEELLAE ARILRQHKSR LETRMQILED HNKQLESQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Members of the dystrophin family of proteins perform a critical role in the maintenance of membrane-associated complexes at points of intercellular contact in vertebrate cells. Dystrophin related protein 2 is predicted to resemble certain short C-terminal isoforms of dystrophin and dystrophin-related protein 1 (DRP1 or utrophin). DRP2 is expressed principally in the brain and spinal cord.
Specifications
DRP2 | |
Polyclonal | |
Unconjugated | |
Drp2 | |
AW495265; DRP2; DRP-2; dystrophin related protein 2; dystrophin-related protein 2; dystrophin-related protein 2 A-form | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1821 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13474 | |
Drp2 | |
Recombinant protein corresponding to Human DRP2. Recombinant protein control fragment (Product #RP-92191). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.