Learn More
Invitrogen™ DRP2 Polyclonal Antibody
Description
Immunogen sequence: MQPMVMQGCP YTLPRCHDWQ AADQFHHSSS LRSTCPHPQV RAAVTSPAPP QDGAGVPCLS LKLLNGSVGA SGPLEPPAMN LCWNEIKKKS HNLRARLEAF SDHSGKLQLP LQEIIDWLSQ KDEELSAQLP LQGDVALVQQ EKETHAAFME EVKSRGPYIY SVLESAQAFL SQHPFEELEE PHSESKDTSP KQRIQNLSRF VWKQATVASE LWEKLTARCV DQHRHIERTL EQLLEIQGAM.
Specifications
Specifications
| Antigen | DRP2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.91 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Drp2 |
| Gene Accession No. | Q05AA6, Q9EPA0 |
| Gene Alias | AW495265; DRP2; DRP-2; dystrophin related protein 2; dystrophin-related protein 2; dystrophin-related protein 2 A-form |
| Gene Symbols | Drp2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.