Learn More
Drucker™ 20 Place Fixed Angle Rotor Accessory
Shop All Drucker Diagnostics Products
Click to view available options
:
100mm
Description
- 20-place tube holder for 100mm tubes
- For use with Drucker 824VES and 853VES model centifuges
Specifications
Specifications
Type | Tube Holder |
For Use With (Equipment) | 653V, 755, 755V and 853VES 20-place fixed angle rotor centrifuge |