Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JUSTRITE Drum Storage Cabinets, 100 gal., Manual Close, with Drum Rollers

Catalog No. 18666763 Shop All Justrite Safety Group Products
Click to view available options
Color:
Blue
Gray
Red
Yellow

Catalog No. 18-666-763

Supplier: JUSTRITE 899163

Only null left
Add to Cart
Add to Cart

For storage of larger quantities of flammable solvents, the heavy-duty Drum Safety Cabinet is available in many styles and sizes.

Built to the same high standards as the smaller Classic Safety Cabinets. An array of drum accessories is available to ensure safety and convenience when storing and handling drums of hazardous materials.

  • 18-gauge (1 mm) double-wall, welded steel with 38 mm of insulating air space for fire resistance.
  • Half-depth shelf adjusts to accommodate a drum with pump or funnel, or stores spare drum accessories.
  • Available in manual or self-close models.
  • Choice of drum rollers to facilitate the movement of heavy drums or removable drum support to keep drums elevated off cabinet sump.
  • Independently fire tested and approved by FM Approvals.
  • High-performance, self-latching doors close easily and securely for maximum protection under fire conditions.
  • U-Loc™ handle comes with a cylinder lock and key set or accepts an optional padlock.
  • Haz-Alert™ reflective warning labels are highly visible under fire conditions.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Designed to meet OSHA and NFPA 30 standards. FM-approved and come with a ten-year warranty

Specifications

Width (Metric) Interior 1422 mm
Width (English) Interior 56 in.
Width (Metric) Exterior 1499 mm
Surface Coating Powder-coat Paint
Weight (Metric) 272.6 kg
Weight (English) 601 lb.
Width (English) Exterior 59 in.
Height (English) Exterior 65 in.
Material Steel (18 ga), Galvanized Steel
No. of Shelves 1
Product Type Vertical Drum Storage Cabinet
Depth (English) Exterior 59 in.
Color Gray
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.