Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drummond™ Digital Microdispensers

Catalog No. 2116920D
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
1000 μL

Includes

Microdispenser 21-169-20D is supplied with 50 glass bores; 21-169-10A is supplied with 7 polypropylene bores; all other models supplied with package of 100 glass bores.

Positive displacement piston eliminates source of errors found with air displacement types

One-hand-operated positive displacement pipetter with digital reading of delivery volume to 0.01, 0.1, or 1.0μL. Five adjustable models cover volume range 0.01 to 1000μL.

  • Adjustable to any volume within capacity rating
  • Easy-to-set micrometer-type barrel for exact setting of volume to be pipetted
  • Accuracy to ±1% at full volume
  • Machined slot in barrel allows visual check for glass bore seating
  • Color-coded calibration mark on bore lines up with plunger at full-capacity setting
  • Borosilicate glass bores with stainless-steel plunger (PTFE tips on 25μL or larger); polypropylene bore on 1000μL size
  • Use with all reagents (except hydrofluoric acid)

Order Info

Microdispenser 21-169-10A converts to a multichannel dispenser with the addition of a stainless-steel manifold.

Specifications

Accuracy ±1%
Color Blue
Number of Channels 1
Type Digital Microdispenser
Volume (Metric) 100 μL
For Use With (Application) Positive lock prevents accidental volume changes
Includes 1 plunger, 2 O-rings, 1 collet, 1 coil spring, 3 PTFE tip, 1 spring guide
Increments 0.1 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.